Gene Transcripts

Gene Symbol
SLC35G5
Entrez Gene ID
83650
Gene Name
solute carrier family 35 member G5
Gene Aliases
AMAC, AMAC1L2
Chromosome Location
Chr. 8: 11330986 - 11332186 on Build GRCh38
UniGene ID
Hs.458397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054028.1 NP_473369.1 1 620
GenBank mRNA AJ291677.1 - 1 620
BC131696.1 - 1 587 40120574

Gene Symbol
SLC35G4
Entrez Gene ID
646000
Gene Name
solute carrier family 35 member G4
Gene Aliases
AMAC1L1, SLC35G4P
Chromosome Location
Chr. 18: 11609558 - 11610612 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282300.1 NP_001269229.1 1 537

Gene Symbol
SLC35G6
Entrez Gene ID
643664
Gene Name
solute carrier family 35 member G6
Gene Aliases
AMAC1L3, TMEM21B
Chromosome Location
Chr. 17: 7481402 - 7483064 on Build GRCh38
UniGene ID
Hs.632234
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102614.1 NP_001096084.1 2 614

View Full Product Details