Gene Transcripts
- Gene Symbol
- CDCA7
- Entrez Gene ID
- 83879
- Gene Name
- cell division cycle associated 7
- Gene Aliases
- ICF3, JPO1
- Chromosome Location
- Chr. 2: 173354017 - 173368992 on Build GRCh38
- UniGene ID
- Hs.470654
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031942.4 |
NP_114148.3
|
4, 5 |
755 |
|
NM_145810.2 |
NP_665809.1
|
3, 4 |
518 |
|
XM_011511957.2 |
XP_011510259.1
|
4, 5 |
1296 |
|
GenBank mRNA |
AK027628.1 |
- |
4, 5 |
727 |
|
AK027642.1 |
- |
3, 4 |
499 |
|
AK075134.1 |
- |
3, 4 |
479 |
|
AK223524.1 |
- |
3, 4 |
523 |
|
AK297097.1 |
- |
2, 3 |
353 |
|
AK297245.1 |
- |
4, 5 |
615 |
|
AK300949.1 |
- |
4, 5 |
584 |
|
AL834186.1 |
- |
3, 4 |
458 |
|
AY029179.1 |
- |
3, 4 |
531 |
|
BC027966.1 |
- |
3, 4 |
484 |
5227112 |
View Full Product Details