Gene Transcripts

Gene Symbol
SLC10A7
Entrez Gene ID
84068
Gene Name
solute carrier family 10 member 7
Gene Aliases
C4orf13, P7
Chromosome Location
Chr. 4: 146253981 - 146522351 on Build GRCh38
UniGene ID
Hs.659209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029998.5 NP_001025169.1 2 377
NM_001300842.2 NP_001287771.1 2 377
NM_001317816.1 NP_001304745.1 2 377
NM_001317817.1 NP_001304746.1 2 377
NM_001317818.1 NP_001304747.1 2 377
NM_032128.4 NP_115504.1 2 377
NR_133924.1 2 377
XM_011532311.2 XP_011530613.1 2 377
XM_011532313.2 XP_011530615.1 1 23
XM_017008689.1 XP_016864178.1 2 377
XM_017008690.1 XP_016864179.1 2 377
XM_017008691.1 XP_016864180.1 2 377
XM_017008692.1 XP_016864181.1 2 757
GenBank mRNA AK291046.1 - 2 245
AL136728.1 - 2 305
AY346324.1 - 2 340
BC023288.1 - 2 328
BC063471.1 - 2 290
BC120970.1 - 2 236
BC120971.1 - 2 236
BC127626.1 - 2 236
BC127627.1 - 2 236
BC148252.1 - 2 236
BC150308.1 - 2 346 8860437
CR933647.1 - 2 371
CR936751.1 - 2 234
DA590625.1 - 2 378
DB205503.1 - 2 377
DQ122860.1 - 2 123
DQ122861.1 - 2 123
DQ871036.1 - 2 123

View Full Product Details