Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032323.2 | NP_115699.1 | 2 | 372 | |
NR_026678.1 | 2 | 397 | |||
GenBank mRNA | AK056816.1 | - | 2 | 337 | |
AK315767.1 | - | 2 | 340 | ||
BC005094.1 | - | 2 | 362 | 3955668 | |
DA678865.1 | - | 2 | 397 | ||
DA880152.1 | - | 2 | 372 |