Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001291963.1 | NP_001278892.1 | 2 | 315 | |
NM_004394.2 | NP_004385.1 | 2 | 315 | ||
GenBank mRNA | AK298670.1 | - | 2 | 268 | |
AK311783.1 | - | 2 | 194 | ||
BC002726.2 | - | 2 | 272 | 3631886 | |
BI823002.1 | - | 2 | 146 | ||
BP265421.1 | - | 2 | 276 | ||
CR542184.1 | - | 2 | 108 | ||
X76105.1 | - | 2 | 267 |