Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318918.1 | NP_001305847.1 | 6 | 1384 | |
NM_032575.2 | NP_115964.2 | 4 | 620 | ||
XM_005255641.4 | XP_005255698.1 | 5 | 685 | ||
GenBank mRNA | AF325914.1 | - | 4 | 620 | |
AK126918.1 | - | 6 | 1384 |