Gene Transcripts

Gene Symbol
SPRYD3
Entrez Gene ID
84926
Gene Name
SPRY domain containing 3
Gene Aliases
Chromosome Location
Chr. 12: 53064316 - 53079420 on Build GRCh38
UniGene ID
Hs.343334
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032840.2 NP_116229.1 2, 3 250
GenBank mRNA AK027706.1 - 2, 3 184
AK074694.1 - 2, 3 216
AK294092.1 - 2, 3 234
BC136312.1 - 2, 3 183 9020299
DA389433.1 - 2, 3 250

View Full Product Details