Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001321027.1 | NP_001307956.1 | 8 | 1223 | |
NM_001354.5 | NP_001345.1 | 11 | 1535 | ||
NM_205845.2 | NP_995317.1 | 10 | 1435 | ||
GenBank mRNA | AB209017.1 | - | 9 | 1190 | |
AK091194.1 | - | 10 | 2789 | ||
AK290304.1 | - | 9 | 1178 | ||
AK296419.1 | - | 8 | 1072 | ||
BC007024.1 | - | 9 | 1144 | 3681970 | |
BC063574.1 | - | 10 | 1402 | 3877666 | |
S68330.1 | - | 9 | 1128 | ||
U05598.1 | - | 9 | 1145 |