Gene Transcripts

Gene Symbol
TRIM5
Entrez Gene ID
85363
Gene Name
tripartite motif containing 5
Gene Aliases
RNF88, TRIM5alpha
Chromosome Location
Chr. 11: 5590906 - 5685109 on Build GRCh38
UniGene ID
Hs.125300
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033034.2 NP_149023.2 2 494
NM_033092.2 NP_149083.2 2 494
NM_033093.2 NP_149084.2 2 346
XM_005253183.2 XP_005253240.1 2 346
XM_005253184.2 XP_005253241.1 2 480
XM_006718358.2 XP_006718421.1 2 480
XM_011520426.2 XP_011518728.1 2 480
XM_011520427.1 XP_011518729.1 2 480
XM_017018460.1 XP_016873949.1 2 480
XM_017018461.1 XP_016873950.1 2 346
XM_017018462.1 XP_016873951.1 2 480
XM_017018463.1 XP_016873952.1 2 346
GenBank mRNA AB362928.1 - 1 191
AF220025.1 - 1 253
AF220026.1 - 253
AF220027.1 - 1 253
AF220028.1 - 1 253
AF220029.1 - 1 253
AK027593.1 - 2 448
AY625000.1 - 1 191
BC020770.1 - 3 617
BC021258.2 - 2 427 4122548
BX647152.1 - 3 383
CN334837.1 - 1 154
DA475412.1 - 2 346
DQ288685.1 - 1 191
JF928461.1 - 1 205
JF928462.1 - 1 205

View Full Product Details