Gene Transcripts

Gene Symbol
TNKS1BP1
Entrez Gene ID
85456
Gene Name
tankyrase 1 binding protein 1
Gene Aliases
TAB182
Chromosome Location
Chr. 11: 57299629 - 57324952 on Build GRCh38
UniGene ID
Hs.530730
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033396.2 NP_203754.2 6 2782
XM_006718725.3 XP_006718788.1 6 2798
XM_011545325.2 XP_011543627.1 6 2795
GenBank mRNA AB051528.2 - 6 2762
AF441771.1 - 5 2924
AK074113.1 - 2 1137
AK124903.1 - 6 2809
AK127871.1 - 4 2205
BC150333.1 - 6 2773 8860508

View Full Product Details