Gene Transcripts

Gene Symbol
EPT1
Entrez Gene ID
85465
Gene Name
ethanolaminephosphotransferase 1
Gene Aliases
SELI, SEPI
Chromosome Location
Chr. 2: 26346086 - 26395891 on Build GRCh38
UniGene ID
Hs.189073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB051511.1 - 2, 3 227
AB621826.1 - 2, 3 220
AK095620.1 - 2, 3 237
BC021229.2 - 2, 3 182 4111329
DA619246.1 - 2, 3 265

View Full Product Details