Gene Transcripts

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
Chromosome Location
Chr. X: 132076986 - 132128022 on Build GRCh38
UniGene ID
Hs.170776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306193.1 NP_001293122.1 6 565
NM_194277.2 NP_919253.1 6 612
XM_017029947.1 XP_016885436.1 6 791
XM_017029948.1 XP_016885437.1 3 253
XM_017029949.1 XP_016885438.1 2 574
GenBank mRNA AK125336.1 - 6 612
AL161984.1 - 2 274
BC114371.1 - 6 579 40079764
FJ717411.1 - 6 389

View Full Product Details