Gene Transcripts

Gene Symbol
YIF1B
Entrez Gene ID
90522
Gene Name
Yip1 interacting factor homolog B, membrane trafficking protein
Gene Aliases
FinGER8
Chromosome Location
Chr. 19: 38303560 - 38315976 on Build GRCh38
UniGene ID
Hs.280741
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039671.2 NP_001034760.1 6 710
NM_001039672.2 NP_001034761.1 6 728
NM_001039673.2 NP_001034762.1 6 719
NM_001145461.1 NP_001138933.1 6 704
NM_001145462.1 NP_001138934.1 6 680
NM_001145463.1 NP_001138935.1 6 719
XM_005259385.3 XP_005259442.1 6 941
XM_017027449.1 XP_016882938.1 6 729
XM_017027450.1 XP_016882939.1 6 727
XM_017027451.1 XP_016882940.1 6 725
GenBank mRNA AF258578.3 - 5 902
AK304479.1 - 3 1066
AL833382.1 - 6 676
AY358965.1 - 5 847
BC007644.1 - 6 667
BC014974.1 - 6 673 4865479
BC091477.1 - 6 702 30344388
BX379501.2 - 6 646
CA487891.1 - 6 662
CA488303.1 - 6 657

View Full Product Details