Gene Transcripts

Gene Symbol
INTS4
Entrez Gene ID
92105
Gene Name
integrator complex subunit 4
Gene Aliases
INT4, MST093
Chromosome Location
Chr. 11: 77874491 - 77994849 on Build GRCh38
UniGene ID
Hs.533723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033547.3 NP_291025.3 5 521
XM_011545352.2 XP_011543654.1 6 690
XM_011545353.2 XP_011543655.1 5 699
XM_017018560.1 XP_016874049.1 5 554
GenBank mRNA AK290866.1 - 5 517
AL834171.1 - 5 464
BC009995.2 - 5 516 4129693
BC015664.2 - 4 414 4554197

View Full Product Details