Gene Transcripts

Gene Symbol
RIPPLY1
Entrez Gene ID
92129
Gene Name
ripply transcriptional repressor 1
Gene Aliases
Chromosome Location
Chr. X: 106900063 - 106903331 on Build GRCh38
UniGene ID
Hs.334726
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001171706.1 NP_001165177.1 2 285
NM_138382.2 NP_612391.1 4 426
GenBank mRNA BC007652.1 - 4 412
BC101998.1 - 4 426
BC105691.1 - 4 426 40023299
BC105692.1 - 4 426 40023298
BC110436.1 - 3 1343 40023301
BC127250.1 - 2 285 40023300
BX106639.1 - 2 321

View Full Product Details