Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001310338.1 | NP_001297267.1 | 6 | 1342 | |
NM_001310339.1 | NP_001297268.1 | 3 | 944 | ||
NM_052865.3 | NP_443097.1 | 5 | 1297 | ||
XM_005260867.2 | XP_005260924.1 | 6 | 1564 | ||
XM_005260870.3 | XP_005260927.1 | 4 | 1112 | ||
XM_006723663.3 | XP_006723726.1 | 5 | 1157 | ||
XM_011529394.1 | XP_011527696.1 | 6 | 1179 | ||
XM_011529395.1 | XP_011527697.1 | 5 | 1057 | ||
XM_017028127.1 | XP_016883616.1 | 5 | 1519 | ||
XM_017028128.1 | XP_016883617.1 | 5 | 1393 | ||
GenBank mRNA | AF306685.1 | - | 5 | 1106 | |
AK027503.1 | - | 5 | 1101 | ||
AK309563.1 | - | 3 | 640 | ||
AK309995.1 | - | 3 | 568 | ||
AK310282.1 | - | 4 | 613 | ||
AK315532.1 | - | 5 | 1133 | ||
BC016869.1 | - | 5 | 1069 | 3850853 |