Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033197.2 | NP_149974.2 | 10 | 1128 | |
GenBank mRNA | AF364078.1 | - | 10 | 1053 | |
AK127971.1 | - | 5 | 595 | ||
AK290243.1 | - | 10 | 1110 | ||
AK313176.1 | - | 10 | 1046 | ||
AY358595.1 | - | 10 | 1040 | ||
BC008429.1 | - | 10 | 1043 | 4291561 |