Gene Transcripts

Gene Symbol
LMTK3
Entrez Gene ID
114783
Gene Name
lemur tyrosine kinase 3
Gene Aliases
LMR3, PPP1R101, TYKLM3
Chromosome Location
Chr. 19: 48485271 - 48513926 on Build GRCh38
UniGene ID
Hs.207426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080434.1 NP_001073903.1 6 625
XM_011526411.2 XP_011524713.1 6 956
XM_011526412.2 XP_011524714.1 6 923
GenBank mRNA AB067470.1 - 6 598

View Full Product Details