Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256124.1 | NP_001243053.1 | 5 | 2034 | |
NM_173547.3 | NP_775818.2 | 6 | 2100 | ||
XM_006721760.3 | XP_006721823.1 | 6 | 2137 | ||
XM_006721761.3 | XP_006721824.1 | 5 | 2071 | ||
XM_017024344.1 | XP_016879833.1 | 4 | 1861 | ||
GenBank mRNA | BC006138.1 | - | 3 | 1209 | |
BC013181.1 | - | 1 | 972 | ||
BC021259.2 | - | 6 | 2068 | 4128330 | |
BC073831.1 | - | 3 | 1297 | ||
BC098412.1 | - | 1024 |