Gene Transcripts

Gene Symbol
TGM7
Entrez Gene ID
116179
Gene Name
transglutaminase 7
Gene Aliases
TGMZ
Chromosome Location
Chr. 15: 43276277 - 43302255 on Build GRCh38
UniGene ID
Hs.164661
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052955.2 NP_443187.1 6 959
XM_017021903.1 XP_016877392.1 6 957
GenBank mRNA AF363393.1 - 6 959

View Full Product Details