Gene Transcripts
- Gene Symbol
- TSEN15
- Entrez Gene ID
- 116461
- Gene Name
- tRNA splicing endonuclease subunit 15
- Gene Aliases
- C1orf19, PCH2F, sen15
- Chromosome Location
- Chr. 1: 184050207 - 184095846 on Build GRCh38
- UniGene ID
- Hs.548197
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127394.3 |
NP_001120866.1
|
1, 2 |
240 |
|
NM_001300764.1 |
NP_001287693.1
|
1, 2 |
240 |
|
NM_001300766.1 |
NP_001287695.1
|
1, 2 |
240 |
|
NM_052965.3 |
NP_443197.1
|
1, 2 |
240 |
|
NR_023349.2 |
|
1, 2 |
240 |
|
NR_125335.1 |
|
1, 2 |
240 |
|
XM_006711148.1 |
XP_006711211.1
|
1, 2 |
189 |
|
XM_011509139.2 |
XP_011507441.1
|
1, 2 |
1684 |
|
XM_017000228.1 |
XP_016855717.1
|
1, 2 |
1684 |
|
XM_017000229.1 |
XP_016855718.1
|
1, 2 |
1684 |
|
XM_017000230.1 |
XP_016855719.1
|
1, 2 |
1684 |
|
GenBank mRNA |
AF288394.1 |
- |
1, 2 |
215 |
|
AK098196.1 |
- |
1, 2 |
138 |
|
AK296655.1 |
- |
1, 2 |
135 |
|
AK307241.1 |
- |
1, 2 |
131 |
|
BC022030.1 |
- |
1, 2 |
185 |
4824025 |
BI821111.1 |
- |
1, 2 |
165 |
|
BM146150.1 |
- |
1, 2 |
242 |
|
BM887891.1 |
- |
1, 2 |
72 |
|
BP326115.1 |
- |
1, 2 |
165 |
|
BU195337.1 |
- |
1, 2 |
212 |
|
View Full Product Details