Gene Transcripts
- Gene Symbol
- BTBD16
- Entrez Gene ID
- 118663
- Gene Name
- BTB domain containing 16
- Gene Aliases
- C10orf87
- Chromosome Location
- Chr. 10: 122270745 - 122338180 on Build GRCh38
- UniGene ID
- Hs.422466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318189.1 |
NP_001305118.1
|
13, 14 |
1433 |
|
NM_144587.3 |
NP_653188.2
|
13, 14 |
1430 |
|
XM_011539239.2 |
XP_011537541.1
|
12, 13 |
1864 |
|
XM_011539240.2 |
XP_011537542.1
|
12, 13 |
1839 |
|
XM_011539241.2 |
XP_011537543.1
|
11, 12 |
1782 |
|
XM_011539242.2 |
XP_011537544.1
|
10, 11 |
1663 |
|
XM_017015636.1 |
XP_016871125.1
|
12, 13 |
2513 |
|
XM_017015637.1 |
XP_016871126.1
|
9, 10 |
2138 |
|
GenBank mRNA |
AK058088.1 |
- |
13, 14 |
1420 |
|
BC109261.1 |
- |
13, 14 |
1404 |
40025404 |
BC109262.1 |
- |
13, 14 |
1404 |
40025414 |
View Full Product Details