Gene Transcripts

Gene Symbol
DOK1
Entrez Gene ID
1796
Gene Name
docking protein 1
Gene Aliases
P62DOK, pp62
Chromosome Location
Chr. 2: 74549020 - 74557551 on Build GRCh38
UniGene ID
Hs.103854
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197260.1 NP_001184189.1 3, 4 555
NM_001318866.1 NP_001305795.1 3, 4 1055
NM_001318867.1 NP_001305796.1 3, 4 1100
NM_001381.4 NP_001372.1 3, 4 1057
GenBank mRNA AF035299.1 - 3, 4 548
AK055944.1 - 3, 4 1057
AK222883.1 - 3, 4 496
AK296154.1 - 3, 4 510
AK314678.1 - 3, 4 477
BC114440.1 - 3, 4 483 40083624
DA382785.1 - 3, 4 555
U70987.1 - 3, 4 484

View Full Product Details