Gene Transcripts

Gene Symbol
KLHL33
Entrez Gene ID
123103
Gene Name
kelch like family member 33
Gene Aliases
Chromosome Location
Chr. 14: 20425852 - 20436509 on Build GRCh38
UniGene ID
Hs.556014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109997.2 NP_001103467.2 4 1677
XM_005267347.4 XP_005267404.1 5 2676
XM_011536450.2 XP_011534752.1 5 2392
GenBank mRNA BC146951.1 - 3 1271
BC146954.1 - 3 1271 9056882
BC151130.1 - 3 1293 9057016

View Full Product Details