Gene Transcripts

Gene Symbol
AFMID
Entrez Gene ID
125061
Gene Name
arylformamidase
Gene Aliases
FKF, KF, KFA
Chromosome Location
Chr. 17: 78185102 - 78207702 on Build GRCh38
UniGene ID
Hs.558614
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010982.4 NP_001010982.2 4, 5 359
NM_001145526.2 NP_001138998.1 4, 5 359
XM_011524326.2 XP_011522628.1 3, 4 291
XM_011524327.2 XP_011522629.1 5, 6 527
XM_011524328.2 XP_011522630.1 5, 6 499
XM_011524329.2 XP_011522631.1 5, 6 541
XM_011524330.2 XP_011522632.1 5, 6 545
XM_011524333.2 XP_011522635.1 4, 5 318
XM_011524336.2 XP_011522638.1 4, 5 318
GenBank mRNA BC132824.1 - 4, 5 359
BX648442.1 - 4, 5 315

View Full Product Details