Gene Transcripts
- Gene Symbol
- LRRC75A-AS1
- Entrez Gene ID
- 125144
- Gene Name
- LRRC75A antisense RNA 1
- Gene Aliases
- C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
- Chromosome Location
- Chr. 17: 16438987 - 16470648 on Build GRCh38
- UniGene ID
- Hs.368934
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027165.1 |
|
2, 3 |
160 |
|
NR_027170.1 |
|
2, 3 |
160 |
|
NR_027667.1 |
|
1, 2 |
426 |
|
NR_045022.1 |
|
2, 3 |
160 |
|
NR_045028.1 |
|
1, 2 |
426 |
|
GenBank mRNA |
BC027986.1 |
- |
1, 2 |
376 |
|
BC040159.1 |
- |
1, 2 |
355 |
5539041 |
BI333700.1 |
- |
1, 2 |
187 |
|
BP870476.1 |
- |
2, 3 |
110 |
|
BX538291.1 |
- |
2, 3 |
111 |
|
DA440445.1 |
- |
2, 3 |
110 |
|
View Full Product Details