Gene Transcripts

Gene Symbol
OR1I1
Entrez Gene ID
126370
Gene Name
olfactory receptor family 1 subfamily I member 1
Gene Aliases
OR19-20, OR1I1P, OR1I1Q
Chromosome Location
Chr. 19: 15087066 - 15088133 on Build GRCh38
UniGene ID
Hs.631610
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004713.1 NP_001004713.1 1 106
GenBank mRNA BC140930.1 - 1 192

View Full Product Details