Gene Transcripts

Gene Symbol
EPYC
Entrez Gene ID
1833
Gene Name
epiphycan
Gene Aliases
DSPG3, PGLB, Pg-Lb, SLRR3B
Chromosome Location
Chr. 12: 90963679 - 91005026 on Build GRCh38
UniGene ID
Hs.435680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004950.4 NP_004941.2 5 613
XM_011538008.1 XP_011536310.1 4 429
GenBank mRNA AK290642.1 - 5 570
BC030958.1 - 5 613 4604494
U59111.1 - 5 566

View Full Product Details