Gene Transcripts
- Gene Symbol
- NCOA7
- Entrez Gene ID
- 135112
- Gene Name
- nuclear receptor coactivator 7
- Gene Aliases
- ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
- Chromosome Location
- Chr. 6: 125780948 - 125932030 on Build GRCh38
- UniGene ID
- Hs.171426
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122842.2 |
NP_001116314.1
|
6, 7 |
951 |
|
NM_001199619.1 |
NP_001186548.1
|
7, 8 |
934 |
|
NM_001199620.1 |
NP_001186549.1
|
8, 9 |
1052 |
|
NM_001199621.1 |
NP_001186550.1
|
5, 6 |
730 |
|
NM_181782.4 |
NP_861447.3
|
6, 7 |
951 |
|
XM_005266822.4 |
XP_005266879.1
|
6, 7 |
981 |
|
XM_006715340.3 |
XP_006715403.1
|
6, 7 |
908 |
|
XM_011535455.2 |
XP_011533757.1
|
6, 7 |
856 |
|
XM_017010269.1 |
XP_016865758.1
|
6, 7 |
667 |
|
XM_017010270.1 |
XP_016865759.1
|
7, 8 |
913 |
|
XM_017010271.1 |
XP_016865760.1
|
6, 7 |
911 |
|
XM_017010272.1 |
XP_016865761.1
|
6, 7 |
764 |
|
XM_017010273.1 |
XP_016865762.1
|
6, 7 |
981 |
|
XM_017010274.1 |
XP_016865763.1
|
6, 7 |
1070 |
|
GenBank mRNA |
AB074157.1 |
- |
6, 7 |
751 |
|
AF493978.1 |
- |
7, 8 |
934 |
|
AK127512.1 |
- |
6, 7 |
761 |
|
AK294558.1 |
- |
5, 6 |
528 |
|
AK307395.1 |
- |
7, 8 |
913 |
|
AL832621.2 |
- |
6, 7 |
744 |
|
AL832628.2 |
- |
6, 7 |
708 |
|
AL834442.1 |
- |
4, 5 |
372 |
|
BC036461.1 |
- |
6, 7 |
779 |
|
BC137094.1 |
- |
8, 9 |
968 |
9021091 |
BC137095.1 |
- |
7, 8 |
850 |
9021092 |
BX537385.1 |
- |
6, 7 |
746 |
|
View Full Product Details