Gene Transcripts

Gene Symbol
E2F1
Entrez Gene ID
1869
Gene Name
E2F transcription factor 1
Gene Aliases
E2F-1, RBAP1, RBBP3, RBP3
Chromosome Location
Chr. 20: 33675486 - 33686404 on Build GRCh38
UniGene ID
Hs.654393
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005225.2 NP_005216.1 3 527
GenBank mRNA BC050369.2 - 3 527
BC058902.1 - 3 458 6055344
M96577.1 - 3 522
S49592.1 - 3 504

View Full Product Details