Gene Transcripts

Gene Symbol
OR13C5
Entrez Gene ID
138799
Gene Name
olfactory receptor family 13 subfamily C member 5
Gene Aliases
OR9-11
Chromosome Location
Chr. 9: 104598457 - 104599413 on Build GRCh38
UniGene ID
Hs.553598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004482.1 NP_001004482.1 1 835
GenBank mRNA BC136842.1 - 1 929
BC136843.1 - 1 929 9020834

View Full Product Details