Gene Transcripts

Gene Symbol
ISM1
Entrez Gene ID
140862
Gene Name
isthmin 1, angiogenesis inhibitor
Gene Aliases
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1
Chromosome Location
Chr. 20: 13221771 - 13326734 on Build GRCh38
UniGene ID
Hs.559353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080826.1 NP_543016.1 3 548
XM_017027680.1 XP_016883169.1 3 548
GenBank mRNA CR982468.1 - 2 329

View Full Product Details