Gene Transcripts
- Gene Symbol
- LAYN
- Entrez Gene ID
- 143903
- Gene Name
- layilin
- Gene Aliases
- Chromosome Location
- Chr. 11: 111540280 - 111563464 on Build GRCh38
- UniGene ID
- Hs.503831
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258390.1 |
NP_001245319.1
|
3, 4 |
736 |
|
NM_001318799.1 |
NP_001305728.1
|
2, 3 |
346 |
|
NM_178834.4 |
NP_849156.1
|
2, 3 |
712 |
|
XM_006718769.3 |
XP_006718832.1
|
2, 3 |
325 |
|
GenBank mRNA |
AK055539.1 |
- |
2, 3 |
713 |
|
AK055654.1 |
- |
2, 3 |
500 |
|
AK293317.1 |
- |
2, 3 |
543 |
|
AK298165.1 |
- |
3, 4 |
371 |
|
AY358554.1 |
- |
3, 4 |
561 |
|
BC025407.1 |
- |
2, 3 |
505 |
4820414 |
DC398135.1 |
- |
2, 3 |
346 |
|
View Full Product Details