Gene Transcripts

Gene Symbol
KRT80
Entrez Gene ID
144501
Gene Name
keratin 80
Gene Aliases
KB20
Chromosome Location
Chr. 12: 52168996 - 52192000 on Build GRCh38
UniGene ID
Hs.140978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081492.1 NP_001074961.1 4 697
NM_182507.2 NP_872313.2 4 697
XM_005268676.3 XP_005268733.1 2 792
GenBank mRNA AJ717743.1 - 4 651
BC065180.1 - 4 697 6573884

View Full Product Details