Gene Transcripts

Gene Symbol
TERB2
Entrez Gene ID
145645
Gene Name
telomere repeat binding bouquet formation protein 2
Gene Aliases
C15orf43
Chromosome Location
Chr. 15: 44956674 - 44979223 on Build GRCh38
UniGene ID
Hs.567701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152448.2 NP_689661.1 6 469
GenBank mRNA BC029537.1 - 6 469

View Full Product Details