Gene Transcripts
- Gene Symbol
- TERB2
- Entrez Gene ID
- 145645
- Gene Name
- telomere repeat binding bouquet formation protein 2
- Gene Aliases
- C15orf43
- Chromosome Location
- Chr. 15: 44956674 - 44979223 on Build GRCh38
- UniGene ID
- Hs.567701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_152448.2 |
NP_689661.1
|
4, 5 |
373 |
|
GenBank mRNA |
BC029537.1 |
- |
4, 5 |
373 |
|
View Full Product Details