Gene Transcripts

Gene Symbol
CMTM4
Entrez Gene ID
146223
Gene Name
CKLF like MARVEL transmembrane domain containing 4
Gene Aliases
CKLFSF4
Chromosome Location
Chr. 16: 66598178 - 66696707 on Build GRCh38
UniGene ID
Hs.643961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178818.2 NP_848933.1 5 829
NM_181521.2 NP_852662.1 4 5532
XM_011522882.1 XP_011521184.1 4 5440
GenBank mRNA AF521889.1 - 5 829
AK126493.1 - 1 3272
CR933620.1 - 1 4441

View Full Product Details