Gene Transcripts

Gene Symbol
SLC35G3
Entrez Gene ID
146861
Gene Name
solute carrier family 35 member G3
Gene Aliases
AMAC1, TMEM21A
Chromosome Location
Chr. 17: 35192520 - 35194393 on Build GRCh38
UniGene ID
Hs.514814
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152462.2 NP_689675.1 1 1813
GenBank mRNA AK097473.1 - 1 1778

View Full Product Details