Gene Transcripts

Gene Symbol
PPM1N
Entrez Gene ID
147699
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1N (putative)
Gene Aliases
Chromosome Location
Chr. 19: 45497964 - 45502694 on Build GRCh38
UniGene ID
Hs.532872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080401.1 NP_001073870.1 5 1493
XM_011526474.2 XP_011524776.1 4 1941
GenBank mRNA AK097444.1 - 4 1896
BC062452.1 - 5 671

View Full Product Details