Gene Transcripts

Gene Symbol
EFNB2
Entrez Gene ID
1948
Gene Name
ephrin B2
Gene Aliases
EPLG5, HTKL, Htk-L, LERK5
Chromosome Location
Chr. 13: 106489731 - 106535040 on Build GRCh38
UniGene ID
Hs.149239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004093.3 NP_004084.1 1 158
GenBank mRNA BC069342.1 - 1 85
BC074856.2 - 1 85 30915225
BC074857.2 - 1 85 30915462
BC105955.1 - 1 103 40034079
BC105956.1 - 1 103 40034078
BC105957.1 - 1 103 40034080
DR004737.1 - 1 166
L38734.1 - 1 113
U16797.1 - 1 89
U81262.1 - 1 89

View Full Product Details