Gene Transcripts

Gene Symbol
ALS2CR11
Entrez Gene ID
151254
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 11
Gene Aliases
Chromosome Location
Chr. 2: 201487421 - 201619182 on Build GRCh38
UniGene ID
Hs.335788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168216.1 NP_001161688.1 5 665
NM_001168217.1 NP_001161689.1 5 665
NM_001168221.1 NP_001161693.1 5 665
NM_152525.5 NP_689738.3 5 665
XM_006712331.3 XP_006712394.1 5 665
XM_006712332.3 XP_006712395.1 5 665
XM_006712333.3 XP_006712396.1 5 665
XM_006712334.3 XP_006712397.1 5 665
XM_006712335.3 XP_006712398.1 5 665
XM_006712336.3 XP_006712399.1 5 665
XM_011510736.2 XP_011509038.1 5 665
GenBank mRNA AK058080.1 - 5 637
AL833429.1 - 5 662
BC030659.2 - 5 658 4838801
BG773299.1 - 5 661
BX648732.1 - 5 658

View Full Product Details