Gene Transcripts

Gene Symbol
HEATR5A
Entrez Gene ID
25938
Gene Name
HEAT repeat containing 5A
Gene Aliases
C14orf125
Chromosome Location
Chr. 14: 31291788 - 31420582 on Build GRCh38
UniGene ID
Hs.744979
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015473.3 NP_056288.2 36 7421
GenBank mRNA AL117511.1 - 1 1414

View Full Product Details