Gene Transcripts

Gene Symbol
KHDC3L
Entrez Gene ID
154288
Gene Name
KH domain containing 3 like, subcortical maternal complex member
Gene Aliases
C6orf221, ECAT1, HYDM2
Chromosome Location
Chr. 6: 73362677 - 73364175 on Build GRCh38
UniGene ID
Hs.128326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017361.2 NP_001017361.1 2 287
GenBank mRNA AB211062.1 - 2 287
BC132844.1 - 2 279 40146866
BC137160.1 - 2 279 9021157

View Full Product Details