Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001420.3 | NP_001411.2 | 3 | 682 | |
NM_032281.2 | NP_115657.2 | 3 | 682 | ||
XM_011527778.2 | XP_011526080.1 | 3 | 685 | ||
GenBank mRNA | AY034002.1 | - | 2 | 271 | |
AY036909.1 | - | 3 | 319 | ||
BC014144.2 | - | 3 | 675 | ||
D26158.1 | - | 3 | 548 | ||
L26405.1 | - | 3 | 448 |