Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_152589.1 | NP_689802.1 | 9 | 919 | |
XM_017018886.1 | XP_016874375.1 | 8 | 859 | ||
XM_017018887.1 | XP_016874376.1 | 9 | 1162 | ||
GenBank mRNA | AK093140.1 | - | 9 | 919 | |
BC038413.2 | - | 8 | 860 | 5170365 |