Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145913.3 | NP_666018.3 | 11 | 1640 | |
XM_017018910.1 | XP_016874399.1 | 11 | 1676 | ||
GenBank mRNA | AF536216.1 | - | 11 | 1613 | |
AK075263.1 | - | 11 | 1298 | ||
AY081220.1 | - | 11 | 1614 | ||
BC110492.2 | - | 11 | 1275 | 40007567 |