Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304787.1 | NP_001291716.1 | 5 | 1643 | |
NM_153360.2 | NP_699191.1 | 4 | 1472 | ||
GenBank mRNA | AK074647.1 | - | 4 | 1427 | |
AK293299.1 | - | 5 | 1602 | ||
BC101758.1 | - | 4 | 1324 | 8069264 | |
BC101760.1 | - | 4 | 1324 | 8069266 |