Gene Transcripts

Gene Symbol
C10orf91
Entrez Gene ID
170393
Gene Name
chromosome 10 open reading frame 91
Gene Aliases
bA432J24.4
Chromosome Location
Chr. 10: 132445210 - 132448321 on Build GRCh38
UniGene ID
Hs.375059
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173541.2 NP_775812.1 3 209
GenBank mRNA BC030794.1 - 3 230
BE909428.1 - 3 209

View Full Product Details