Gene Transcripts

Gene Symbol
SLC29A1
Entrez Gene ID
2030
Gene Name
solute carrier family 29 member 1 (Augustine blood group)
Gene Aliases
ENT1
Chromosome Location
Chr. 6: 44219505 - 44234151 on Build GRCh38
UniGene ID
Hs.25450
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078175.2 NP_001071643.1 8, 9 987
NM_001078177.1 NP_001071645.1 8, 9 1182
NM_001304462.1 NP_001291391.1 8, 9 1120
NM_001304463.1 NP_001291392.1 7, 8 880
NM_001304465.1 NP_001291394.1 7, 8 1023
NM_001304466.1 NP_001291395.1 7, 8 1020
XM_005248876.4 XP_005248933.1 7, 8 845
XM_005248878.3 XP_005248935.1 8, 9 1017
XM_005248879.3 XP_005248936.1 8, 9 951
XM_005248880.3 XP_005248937.1 8, 9 928
XM_005248881.3 XP_005248938.1 7, 8 1025
XM_005248882.3 XP_005248939.1 7, 8 968
XM_011514341.2 XP_011512643.1 8, 9 1041
GenBank mRNA AF079117.1 - 7, 8 793
AK090491.1 - 8, 9 989
AK090615.1 - 8, 9 1008
AX775997.1 - 7, 8 869
BC001382.2 - 7, 8 793 3051441
BC008954.2 - 8, 9 866 3010092
U81375.1 - 7, 8 869

View Full Product Details