Gene Transcripts

Gene Symbol
GKN2
Entrez Gene ID
200504
Gene Name
gastrokine 2
Gene Aliases
BRICD1B, GDDR, PRO813, TFIZ1, VLTI465
Chromosome Location
Chr. 2: 68945232 - 68952970 on Build GRCh38
UniGene ID
Hs.16757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182536.2 NP_872342.2 2 150
GenBank mRNA AF494509.2 - 2 150
AJ966788.1 - 2 71
AY358664.1 - 2 125
AY943908.1 - 2 73

View Full Product Details